Sodium Dodecyl Sulfate (SDS), White Powder, Electrophoresis, Fisher BioReagents™
Sodium Dodecyl Sulfate (SDS), White Powder, Electrophoresis, Fisher BioReagents™
Sodium Dodecyl Sulfate (SDS), White Powder, Electrophoresis, Fisher BioReagents™
Fisher BioReagents™

Sodium Dodecyl Sulfate (SDS), White Powder, Electrophoresis, Fisher BioReagents™

For protein extraction procedures and SDS polyacrylamide gels | CAS: 151-21-3 | C12H25NaO4S | 288.38 g/mol
Quantity:
100 g
500 g
Packaging:
View all
Poly Bottle
Catalog number BP166-500
Price (JPY)
-
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
Absorbance0.1 max. (0.1% solution) at 280nm, 0.4 max. (0.1% solution) at 230nm
Assay Percent Notes(as Fatty Alcohol Sulfate) >=99%
Assay Percent Range(as Fatty Alcohol Sulfate) >=99%
CAS151-21-3
CAS7647-14-5
View more
For protein extraction procedures and SDS polyacrylamide gels
  • DNase and RNase-free detergent for protein extraction procedures and polyacrylamide gel electrophoresis
  • SDS is a commonly used detergent in protein purification and polyacrylamide gel electrophoresis (PAGE)
  • SDS denatures proteins by disrupting the non-covalent bonds of the protein
  • Functionally tested for use in electrophoresis
  • Intended for research use only; not for human or animal diagnostic purposes