Phenol, Saturated (pH 6.6/7.9, Liq.), Fisher BioReagents™
Phenol, Saturated (pH 6.6/7.9, Liq.), Fisher BioReagents™
Phenol, Saturated (pH 6.6/7.9, Liq.), Fisher BioReagents™
Fisher BioReagents™

Phenol, Saturated (pH 6.6/7.9, Liq.), Fisher BioReagents™

Catalog NumberQuantityPackaging
BP1750I-400400 mLCoated Amber Glass/Poison Pack
Catalog number BP1750I-400
Price (JPY)
-
Quantity:
400 mL
Packaging:
Coated Amber Glass/Poison Pack
Request bulk or custom format
Specifications
Assay Percent Range≥99.0
CAS108-95-2
CAS6381-92-6
CAS77-86-1
ChEBICHEBI:15882
View more
Phenol, Saturated (pH 6.6/7.9, Liq.)
Phenol saturated with Tris buffer is used in DNA extraction procedures.