Chemical Name or MaterialPhenol/Chloroform/Isoamyl Alcohol
View more
For extraction of proteins from crude nucleic acid preparations
This mixture is used for extracting protein from crude nucleic acid preparations. Solution comes with a bottle of tris that should be used to equilibrate the pH.