Phenol/Chloroform/Isoamyl Alcohol (125:24:1 Mixture, pH 4.3, Liq.), Fisher BioReagents™
Phenol/Chloroform/Isoamyl Alcohol (125:24:1 Mixture, pH 4.3, Liq.), Fisher BioReagents™
Phenol/Chloroform/Isoamyl Alcohol (125:24:1 Mixture, pH 4.3, Liq.), Fisher BioReagents™
Fisher BioReagents™

Phenol/Chloroform/Isoamyl Alcohol (125:24:1 Mixture, pH 4.3, Liq.), Fisher BioReagents™

Catalog NumberQuantityPackaging
BP1754I-100100 mLCoated Amber Glass/Poison Pack
Catalog number BP1754I-100
Price (JPY)
-
Quantity:
100 mL
Packaging:
Coated Amber Glass/Poison Pack
Request bulk or custom format
Specifications
CAS123-51-3
ChEBICHEBI:15837
ChemAlert Storage SymbolWhite
Chemical Name or MaterialPhenol/Chloroform/Isoamyl Alcohol
DOT InformationDOT Class 6.1, : Poison
View more
Phenol/Chloroform/Isoamyl Alcohol (125:24:1 Mixture, pH 4.3, Liq.)
This mixture is used to purify total RNA from mixtures containing RNA, DNA, and protein.