Ampicillin Sodium Salt (Crystalline Powder), Fisher BioReagents™
Ampicillin Sodium Salt (Crystalline Powder), Fisher BioReagents™
Ampicillin Sodium Salt (Crystalline Powder), Fisher BioReagents™
Fisher BioReagents™

Ampicillin Sodium Salt (Crystalline Powder), Fisher BioReagents™

For selection for ampicillin resistance in mutated and transformed cells | CAS: 69-52-3 | C16H21N3NaO4S | 374.411 g/mol
Quantity:
5 g
25 g
Packaging:
Poly Bottle
Catalog number BP1760-25
Price (JPY)
-
Quantity:
25 g
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
CAS69-52-3
ChemAlert Storage SymbolBlue
Chemical Name or MaterialAmpicillin Sodium Salt
Formula Weight371.39
Health Hazard 2WARNING!
View more
For selection for ampicillin resistance in mutated and transformed cells
For selection for ampicillin resistance in mutated and transformed cells. Antimicrobial spectrum: Gram-negative and Gram-positive bacteria.