Ammonium Persulfate (Colorless-to-White Crystals/Electrophoresis), Fisher BioReagents™
Ammonium Persulfate (Colorless-to-White Crystals/Electrophoresis), Fisher BioReagents™
Ammonium Persulfate (Colorless-to-White Crystals/Electrophoresis), Fisher BioReagents™
Fisher BioReagents™

Ammonium Persulfate (Colorless-to-White Crystals/Electrophoresis), Fisher BioReagents™

CAS: 7727-54-0 | H8N2O8S2 | 228.19 g/mol
Quantity:
25 g
100 g
Packaging:
Glass Bottle
Catalog number BP179-100
Price (JPY)
-
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Assay Percent Range≥98.0 %
CAS7727-54-0
ChemAlert Storage SymbolYellow
Chemical Name or MaterialAmmonium Persulfate
ColorWhite
View more
Ammonium Persulfate (Colorless-to-White Crystals/Electrophoresis)
Electrophoresis-grade Ammonium Persulfate is an initiator of acrylamide polymerization for the preparation of polyacrylamide electrophoresis gels.