Water, DNA Grade, DNASE, Protease free, Fisher BioReagents
Water, DNA Grade, DNASE, Protease free, Fisher BioReagents
Water, DNA Grade, DNASE, Protease free, Fisher BioReagents
Fisher BioReagents™

Water, DNA Grade, DNASE, Protease free, Fisher BioReagents

CAS: 7732-18-5 | H2O | 18.015 g/mol
Catalog NumberQuantityPackaging
BP2470-11 LNalgene Poly Bottle
Catalog number BP2470-1
Price (JPY)
-
Quantity:
1 L
Packaging:
Nalgene Poly Bottle
Nalgene Poly Bottle; 1L
DNA-grade Water is DNase- and Protease-free and is suitable for all DNA work.
Specifications
Boiling Point100°C
CAS7732-18-5
ChEBICHEBI:15377
ChemAlert Storage SymbolGray
Chemical Name or MaterialWater
ColorColorless
DNaseDNase free
Density1.000g/cm³
Filtered ThroughFiltered through a 0.2-micron filter and autoclaved.
Formula Weight18.02
IUPAC Nameoxidane
InChI KeyXLYOFNOQVPJJNP-UHFFFAOYSA-N
Melting Point0°C
Merck Index15, 10237
Molecular FormulaH2O
Name NoteFor DNA Work, DNASE, Protease free
PackagingNalgene Poly Bottle
Physical FormLiquid
PubChem CID962
SMILESO
Synonymdihydrogen oxide, dihydrogen monoxide
Vapor Pressure17.5mmHg at 20°C
GradeDNA
Molecular Weight (g/mol)18.015
Quantity1 L
pH7
Unit SizeEach