Ionomycin (Calcium Salt), Fisher BioReagents™
Ionomycin (Calcium Salt), Fisher BioReagents™
Ionomycin (Calcium Salt), Fisher BioReagents™
Fisher BioReagents™

Ionomycin (Calcium Salt), Fisher BioReagents™

Catalog NumberQuantityPackaging
BP2527-11 mgPoly Micro Tube
Catalog number BP2527-1
Price (JPY)
-
Quantity:
1 mg
Packaging:
Poly Micro Tube
Request bulk or custom format
Specifications
Assay Percent Range≥98.0 %
CAS56092-82-1
ChemAlert Storage SymbolGray
Chemical Name or MaterialIonomycin, Calcium Salt
Formula Weight747.1
View more
Ionomycin (Calcium Salt)
  • Ionomycin functions as a calcium ionophore or ion carrier and is commonly used to study Ca2+ transport across biological membranes
  • Ionomycin, in conjunction with PMA, can also be used to induce cytokine production in many cell types
  • Store product at 2-8°C and protected from light
  • Intended for research use only