Mitomycin C, Fisher BioReagents™
Mitomycin C, Fisher BioReagents™
Mitomycin C, Fisher BioReagents™
Fisher BioReagents™

Mitomycin C, Fisher BioReagents™

Catalog NumberQuantityPackaging
BP2531-1010 mgAmber Glass
Catalog number BP2531-10
Price (JPY)
-
Quantity:
10 mg
Packaging:
Amber Glass
Request bulk or custom format
Specifications
Assay Percent Range≥98.0 %
CAS50-07-7
ChEBICHEBI:27504
ChemAlert Storage SymbolBlue
Chemical Name or MaterialMitomycin C
View more
Mitomycin C
Inhibitor of DNA synthesis, nuclear division, and cancer cells. Antibacterial spectrum: Gram positive, gram negative, acid-fast bacilli.

Recommended Storage: 4°C. Keep containers tightly closed in a dry, cool and well-ventilated place. Keep refrigerated.