<i>ex</i>ACTG<i>ene</i>&trade; DNA Ladders
<i>ex</i>ACTG<i>ene</i>&trade; DNA Ladders
<i>ex</i>ACTG<i>ene</i>&trade; DNA Ladders
Fisher BioReagents™

exACTGene™ DNA Ladders

Ready-to-use DNA size standards are stable at room temperature—no need to thaw and refreeze
Catalog NumberFor Use With (Application)No. of Bands
BP2574-100Fast run times, small size DNA11
BP2571-100PCR size confirmation12
Catalog number BP2574-100
Price (JPY)
-
For Use With (Application):
Fast run times, small size DNA
No. of Bands:
11
Ready-to-use DNA size standards are stable at room temperature—no need to thaw and refreeze

  • Specially engineered to provide blunt-end DNA of precise length for better band quality than classic markers
  • Ideal for size assessment and quantitative determination of DNA mass
  • Contain loading dye to reduce pipetting steps and save time
  • Have two or three high intensity reference bands for better sizing
  • Provided in sufficient quantity to load 100 lanes

Specifications
For Use With (Application)Fast run times, small size DNA
FormLiquid
FormulationPremixed with loading dye
No. of Bands11
Quantity100 Lanes
Storage RequirementsStable at room temperature for two years.
Sufficient For100 Lanes
ColorColorless
Product TypeDNA Molecular Weight Marker
Unit SizeEach