Doxycycline Hydrochloride (Yellow Powder), Fisher BioReagents™
Doxycycline Hydrochloride (Yellow Powder), Fisher BioReagents™
Doxycycline Hydrochloride (Yellow Powder), Fisher BioReagents™
Fisher BioReagents™

Doxycycline Hydrochloride (Yellow Powder), Fisher BioReagents™

Broad spectrum antibiotic
Catalog NumberQuantityPackaging
BP2653-55 gAmber Glass
Catalog number BP2653-5
Price (JPY)
-
Quantity:
5 g
Packaging:
Amber Glass
Request bulk or custom format
Specifications
AbsorbanceInclusive between 300 at 349nm
Assay Percent Range95%
CAS10592-13-9
ChemAlert Storage SymbolGray
Chemical Name or MaterialDoxycycline Hydrochloride
View more
Broad spectrum antibiotic
Broad spectrum antibiotic. Broad spectrum inhibitor of matrix metalloproteinases in vivo.