Rifampicin, Fisher BioReagents™
Rifampicin, Fisher BioReagents™
Rifampicin, Fisher BioReagents™
Fisher BioReagents™

Rifampicin, Fisher BioReagents™

Catalog NumberQuantityPackaging
BP2679-11 gAmber Glass
Catalog number BP2679-1
Price (JPY)
-
Quantity:
1 g
Packaging:
Amber Glass
Request bulk or custom format
Specifications
Assay Percent Range97%
CAS13292-46-1
ChemAlert Storage SymbolGray
Chemical Name or MaterialRifampicin
Formula Weight822.96
View more
Rifampicin
Rifampicin is a rifamycin which specifically inhibits DNA-dependent bacterial RNA Polymerase. Mammalian RNA polymerase is not affected.