Ethanol, Absolute (200 Proof), Molecular Biology Grade, Fisher BioReagents™
Ethanol, Absolute (200 Proof), Molecular Biology Grade, Fisher BioReagents™
Ethanol, Absolute (200 Proof), Molecular Biology Grade, Fisher BioReagents™
Fisher BioReagents™

Ethanol, Absolute (200 Proof), Molecular Biology Grade, Fisher BioReagents™

Meets ACS Specifications
Catalog NumberQuantity
BP2818-100100 mL
Catalog number BP2818-100
Price (JPY)
-
Quantity:
100 mL
Request bulk or custom format
Specifications
Assay Percent RangeAssay: ≥99.5% (v/v)
Boiling Point78°C
CAS64-17-5
ChEBICHEBI:16236
ChemAlert Storage SymbolRed
View more
Meets ACS Specifications
  • Meets ACS Specifications
  • For nucleic acid purification and precipitation
  • Ultrapure molecular biology grade ethanol is used for the purification and precipitation of biomolecules such as nucleic acids and proteins
  • It is also used in histology to prepare staining and destaining reagents and for dehydrating tissues prior to embedding