PBS Tablets, Phosphate Buffered Saline, Fisher BioReagents™
PBS Tablets, Phosphate Buffered Saline, Fisher BioReagents™
PBS Tablets, Phosphate Buffered Saline, Fisher BioReagents™
Fisher BioReagents™

PBS Tablets, Phosphate Buffered Saline, Fisher BioReagents™

Phosphate Buffered Saline Tablets, CAS Number-7647-14-5, 7447-40-7, 100 tablets, Clear, Gray, White, 5/8 in., 600 SEC max., CAUTION!
Catalog NumberQuantity
BP2944-100100 Tablets
Catalog number BP2944-100
Price (JPY)
-
Quantity:
100 Tablets
Phosphate Buffered Saline Tablets, CAS Number-7647-14-5, 7447-40-7, 100 tablets, Clear, Gray, White, 5/8 in., 600 SEC max., CAUTION!
  • Save time and simplify your buffer preparation step by using Fisher BioReagents PBS tablets
  • Eliminate the hassle of weighing and mixing individual components
  • One tablet dissolved in 200mL water yields buffer containing 0.01M phosphate, 0.0027M KCl, and 0.137M NaCl, pH 7.4 at 25°C
  • Bottle contains 100 tablets
  • Intended for laboratory research use only
Specifications
ColorWhite
PackagingPoly Bottle
Physical FormSolid
Quantity100 Tablets
Solution TypePhosphate Buffered Saline
pH7.2 to 7.6
Unit SizeEach