Puromycin Dihydrochloride, Fisher BioReagents™
Puromycin Dihydrochloride, Fisher BioReagents™
Puromycin Dihydrochloride, Fisher BioReagents™
Fisher BioReagents™

Puromycin Dihydrochloride, Fisher BioReagents™

Inhibits protein synthesis | CAS: 58-58-2 | C22H31Cl2N7O5 | 544.43 g/mol
Catalog NumberQuantity
BP2956-100100 mg
Catalog number BP2956-100
Price (JPY)
-
Quantity:
100 mg
Inhibits protein synthesis
Specifications
Assay Percent Range> 98 %
CAS58-58-2
Chemical Name or MaterialPuromycin Dihydrochloride
Formula Weight544.44
Health Hazard 2WARNING!
Health Hazard 3Emergency Overview
Harmful if swallowed. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Call a physician or Poison Control Center immediately.

NFPA

Health:2
Flammability:0
Instability:0
IUPAC Name(2S)-2-amino-N-[(2S,3S,4S,5R)-5-[6-(dimethylamino)purin-9-yl]-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]-3-(4-methoxyphenyl)propanamide;dihydrochloride
InChI KeyMKSVFGKWZLUTTO-MLYJQVMKNA-N
MDL NumberMFCD00150080
Melting Point168°C
Merck Index15, 8054
Molecular FormulaC22H31Cl2N7O5
PackagingClear Glass
PubChem CID131632508
SMILESCl.Cl.COC1=CC=C(C[C@H](N)C(=O)N[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O)N2C=NC3=C(N=CN=C23)N(C)C)C=C1
SourceStreptomyces alboniger
Water<12.0
ColorBeige
Molecular Weight (g/mol)544.43
Percent Purity98% min.
Physical FormSolid
Quantity100 mg
TypePuromycin Dihydrochloride
Unit SizeEach