HEPES (Fine White Crystals/Molecular Biology), Fisher BioReagents™
HEPES (Fine White Crystals/Molecular Biology), Fisher BioReagents™
HEPES (Fine White Crystals/Molecular Biology), Fisher BioReagents™
Fisher BioReagents™

HEPES (Fine White Crystals/Molecular Biology), Fisher BioReagents™

Commonly used buffering agent | CAS: 7365-45-9 | C8H18N2O4S | 238.30 g/mol
Quantity:
500 g
1 kg
5 kg
Packaging:
View all
Poly Bottle
Catalog number BP310-500
Price (JPY)
-
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Chemical Identifiers
CAS7365-45-9
Specifications
Absorbance0.01 max. (0.1M solution) at 280nm
Assay Percent Range≥99 %
CAS7365-45-9
ChEBICHEBI:42334
ChemAlert Storage SymbolGray
View more
Commonly used buffering agent
HEPES is commonly used as a buffering agent.