Sodium Pyruvate (White Powder), Fisher BioReagents
Sodium Pyruvate (White Powder), Fisher BioReagents
Sodium Pyruvate (White Powder), Fisher BioReagents
Fisher BioReagents™

Sodium Pyruvate (White Powder), Fisher BioReagents

CAS: 113-24-6 | C3H3NaO3 | 110.044 g/mol
Catalog NumberQuantityPackaging
BP356-100100 gGlass Bottle
Catalog number BP356-100
Price (JPY)
-
Quantity:
100 g
Packaging:
Glass Bottle
Glass Bottle; 100g
This sodium salt is used in buffers for molecular biology and cell culture applications.
Specifications
Assay Percent Range≥99 %
CAS113-24-6
ChEBICHEBI:50144
ChemAlert Storage SymbolGray
Chemical Name or MaterialSodium Pyruvate
ColorWhite
Formula Weight110.04
IUPAC Namesodium;2-oxopropanoate
InChI KeyDAEPDZWVDSPTHF-UHFFFAOYSA-M
Linear FormulaCH3COCO2Na
Melting Point300°C
Molecular FormulaC3H3NaO3
PackagingGlass Bottle
PubChem CID23662274
Recommended StorageRT
SMILESCC(=O)C(=O)[O-].[Na+]
Synonymsodium pyruvate, pyruvic acid sodium salt, sodium 2-oxopropanoate, pyruvic acid, sodium salt, propanoic acid, 2-oxo-, sodium salt, sodium alpha-ketopropionate, pyruvate sodium, unii-pod38aif08, 2-oxopropanoic acid sodium salt, pod38aif08
Molecular Weight (g/mol)110.044
Percent Purity≥99%
Physical FormPowder or Solid
Quantity100 g
pH5.5 to 6.5
Unit SizeEach