Ammonium Phosphate Dibasic (Colorless Crystals), Fisher BioReagents
Ammonium Phosphate Dibasic (Colorless Crystals), Fisher BioReagents
Ammonium Phosphate Dibasic (Colorless Crystals), Fisher BioReagents
Fisher BioReagents™

Ammonium Phosphate Dibasic (Colorless Crystals), Fisher BioReagents

CAS: 7783-28-0 | H9N2O4P | 132.06 g/mol
Catalog NumberQuantityPackaging
BP361-500500 gPoly Bottle
Catalog number BP361-500
Price (JPY)
-
Quantity:
500 g
Packaging:
Poly Bottle
Poly Bottle; 500g
Ammonium Phosphate Dibasic is suitable for use in many biological applications.
Specifications
Assay Percent Range98.0 to 101.0 %
CAS7783-28-0
ChEBICHEBI:63051
ChemAlert Storage SymbolGray
Chemical Name or MaterialAmmonium Phosphate Dibasic
ColorWhite
Density1.619g/cm³
Formula Weight132.06
IUPAC Namephosphoric acid diamine
IdentificationPass Test
InChI KeyMNNHAPBLZZVQHP-UHFFFAOYSA-N
Linear Formula(NH4)2HPO4
MDL NumberMFCD00010891
Merck Index15, 540
Molecular FormulaH9N2O4P
PackagingPoly Bottle
PubChem CID24540
Recommended StorageRT
SMILESN.N.OP(O)(O)=O
Synonymdiammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate
Molecular Weight (g/mol)132.06
Percent Purity98.0 to 101.0%
Physical FormCrystalline Solid
Quantity500 g
pH7.9 to 8.3
Unit SizeEach