Polyvinylpyrrolidone, Fisher BioReagents™
Polyvinylpyrrolidone, Fisher BioReagents™
Polyvinylpyrrolidone, Fisher BioReagents™
Fisher BioReagents™

Polyvinylpyrrolidone, Fisher BioReagents™

Polyvinylpyrrolidone, >-95%, C6H9NO, CAS Number-9003-39-8, n-vinyl-2-pyrrolidinone, 2-pyrrolidinone, 1-ethenyl, n-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, 1-vinylpyrrolidone, 1-vinyl-2-pyrrolidinone, 1-vinyl-2-pyrrolidone, n-vinylpyrrolidone, n-vinylpyrrolidinone, vinylpyrrolidone, 500g
Catalog NumberQuantityPackaging
BP431-500500 gPoly Bottle
Catalog number BP431-500
Price (JPY)
-
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
Ash0.05% max.
Assay Percent Notes(dry basis)≥95%
Assay Percent Range≥95 %
CAS9003-39-8
ChEBICHEBI:82551
View more

Polyvinylpyrrolidone, >-95%, C6H9NO, CAS Number-9003-39-8, n-vinyl-2-pyrrolidinone, 2-pyrrolidinone, 1-ethenyl, n-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, 1-vinylpyrrolidone, 1-vinyl-2-pyrrolidinone, 1-vinyl-2-pyrrolidone, n-vinylpyrrolidone, n-vinylpyrrolidinone, vinylpyrrolidone, 500g

Polyvinyl Pyrrolidone is a component of Denhardt's Reagent, commonly used in nucleic acid hybridization procedures.