Calcium Chloride Dihydrate (White Crystals to Powder), Fisher BioReagents™
Calcium Chloride Dihydrate (White Crystals to Powder), Fisher BioReagents™
Calcium Chloride Dihydrate (White Crystals to Powder), Fisher BioReagents™
Fisher BioReagents™

Calcium Chloride Dihydrate (White Crystals to Powder), Fisher BioReagents™

Catalog NumberQuantityPackaging
BP510-500500 gPoly Bottle
Catalog number BP510-500
Price (JPY)
-
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
Assay Percent Range99.0 to 105.0 %
CAS10035-04-8
CAS10043-52-4
ChemAlert Storage SymbolGray
Chemical Name or MaterialCalcium Chloride Dihydrate
View more
Calcium Chloride Dihydrate (White Crystals to Powder)
Calcium Chloride is suitable for use in the preparation of buffer solutions.