Ficoll 400 (White to Off-White Powder/Molecular Biology), Fisher BioReagents™
Ficoll 400 (White to Off-White Powder/Molecular Biology), Fisher BioReagents™
Ficoll 400 (White to Off-White Powder/Molecular Biology), Fisher BioReagents™
Fisher BioReagents™

Ficoll 400 (White to Off-White Powder/Molecular Biology), Fisher BioReagents™

Catalog NumberQuantityPackaging
BP525-2525 gPoly Bottle
Catalog number BP525-25
Price (JPY)
-
Quantity:
25 g
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
CAS26873-85-8
ChemAlert Storage SymbolGray
Chemical Name or MaterialFicoll™ 400
ColorWhite
Formula Weight≈400,000
View more
Ficoll 400 (White to Off-White Powder/Molecular Biology)

Ficoll 400 is used as a component of Denhardt's Reagent and in gel-loading buffers for DNA gels. It is also used in forming density gradients to separate cells and subcellular components.