Calcium Chloride Dihydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., USP, Fisher Chemical™
Calcium Chloride Dihydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., USP, Fisher Chemical™
Calcium Chloride Dihydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., USP, Fisher Chemical™
Calcium Chloride Dihydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., USP, Fisher Chemical™
Fisher Chemical™

Calcium Chloride Dihydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., USP, Fisher Chemical™

Quantity:
250 g
500 g
Packaging:
Plastic powder jar
Catalog number C-1500-53
also known as C/1500/53
Price (JPY)
-
Contact Us ›
Quantity:
500 g
Packaging:
Plastic powder jar
Request bulk or custom format
Specifications
CAS10035-04-8
ColorBeige to White
Formula Weight147.02g/mol
IUPAC Namecalcium dihydrate dichloride
InChI KeyLLSDKQJKOVVTOJ-UHFFFAOYSA-L
View more
Calcium Chloride Dihydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., USP