Calcium Hydroxide, Certified AR for Analysis, Fisher Chemical™
Calcium Hydroxide, Certified AR for Analysis, Fisher Chemical™
Calcium Hydroxide, Certified AR for Analysis, Fisher Chemical™
Fisher Chemical™

Calcium Hydroxide, Certified AR for Analysis, Fisher Chemical™

Catalog NumberQuantity
C-1642-53
also known as C/1642/53
500 g
Catalog number C-1642-53
also known as C/1642/53
Price (JPY)
-
Quantity:
500 g
Request bulk or custom format
Specifications
Boiling Point2850°C
CAS1305-62-0
ColorWhite
Formula Weight74.09g/mol
IUPAC Namecalcium dihydroxide
View more
Calcium Hydroxide, Certified AR for Analysis