Chloroform, 99.8+%, Certified AR for Analysis, Stabilised with Amylene, Fisher Chemical™
Chloroform, 99.8+%, Certified AR for Analysis, Stabilised with Amylene, Fisher Chemical™
Chloroform, 99.8+%, Certified AR for Analysis, Stabilised with Amylene, Fisher Chemical™
Chloroform, 99.8+%, Certified AR for Analysis, Stabilised with Amylene, Fisher Chemical™
Fisher Chemical™

Chloroform, 99.8+%, Certified AR for Analysis, Stabilised with Amylene, Fisher Chemical™

CHCl3, CAS Number-67-66-3, 2.5L, 61 deg.C, Colorless, 119.38g/mol, -63 deg.C, 826, Plastic Coated Glass bottle, Liquid, 213mbar at 20 deg.C, 0.56 mPaS at 20 deg.C | CAS: 67-66-3 | CHCl3 | 119.37 g/mol
Quantity:
1 L
2.5 L
Packaging:
Plastic-coated Glass Bottle
Amber Glass Bottle
Catalog number C-4960-PB17
also known as C/4960/PB17
Price (JPY)
-
Contact Us ›
Quantity:
2.5 L
Packaging:
Plastic-coated Glass Bottle
Request bulk or custom format
Specifications
Boiling Point61°C
CAS67-66-3
ColorColorless
Formula Weight119.38g/mol
IUPAC Nametrichloromethane
View more
CHCl3, CAS Number-67-66-3, 2.5L, 61 deg.C, Colorless, 119.38g/mol, -63 deg.C, 826, Plastic Coated Glass bottle, Liquid, 213mbar at 20 deg.C, 0.56 mPaS at 20 deg.C