Chloroform, Extra Dry, for Synthesis, Stabilised with Amylene, Fisher Chemical™
Chloroform, Extra Dry, for Synthesis, Stabilised with Amylene, Fisher Chemical™
Chloroform, Extra Dry, for Synthesis, Stabilised with Amylene, Fisher Chemical™
Fisher Chemical™

Chloroform, Extra Dry, for Synthesis, Stabilised with Amylene, Fisher Chemical™

Catalog NumberQuantity
C-4962-15
also known as C/4962/15
1 L
Catalog number C-4962-15
also known as C/4962/15
Price (JPY)
-
Quantity:
1 L
Request bulk or custom format
Specifications
Boiling Point61°C
CAS67-66-3
ColorColorless
Formula Weight119.38g/mol
IUPAC Nametrichloromethane
View more
Chloroform, Extra Dry, for Synthesis, Stabilised with Amylene