Copper, Technical, Turnings, Metal, SLR, Fisher Chemical™
Copper, Technical, Turnings, Metal, SLR, Fisher Chemical™
Copper, Technical, Turnings, Metal, SLR, Fisher Chemical™
Copper, Technical, Turnings, Metal, SLR, Fisher Chemical™
Fisher Chemical™

Copper, Technical, Turnings, Metal, SLR, Fisher Chemical™

Cu, CAS Number-7440-50-8, 500g, 2595 deg.C, Brown, 63.54g/mol, 1083 deg.C, 10965, Plastic powder jar, Solid
Catalog NumberQuantity
C-7160-53
also known as C/7160/53
500 g
Catalog number C-7160-53
also known as C/7160/53
Price (JPY)
-
Contact Us ›
Quantity:
500 g
Request bulk or custom format
Specifications
Boiling Point2595°C
CAS7440-50-8
ColorBrown
Formula Weight63.54g/mol
IUPAC Namecopper
View more
Cu, CAS Number-7440-50-8, 500g, 2595 deg.C, Brown, 63.54g/mol, 1083 deg.C, 10965, Plastic powder jar, Solid