Product Image
Gibco™
Gibco™ BASIC PBS, pH 7.4
Catalog number C10010500BT
Price (JPY)
-
Phosphate-Buffered Saline (PBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. PBS is formulated without calcium and magnesium for rinsing chelators from the culture before cell dissociation.
This PBS is manufactured as follows:
Without
• Calcium
• Magnesium
• Phenol Red
Gibco® 'BASIC' products offer value-minded scientists liquid cell culture media and buffer solutions at a significant price savings. As part of Life Technologies’ commitment to quality, this select range of Research Use Only products is continually tested to ensure performance and customer satisfaction.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Unit Size500 mL
Contents & Storage
Storage conditions: 2-8° C. Protect from lightShipping conditions: AmbientShelf life: 12 months from date of manufacture