Product Image
Gibco™
Gibco™ BASIC DMEM, High Glucose, no Glutamine
Catalog number C11960500BT
Price (JPY)
-
Dulbecco's Modified Eagle Medium (DMEM) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs, and smooth muscle cells, as well as cell lines such as HeLa, 293, Cos-7, and PC-12.
This DMEM is modified as follows:
WithWithout
• High Glucose• L-glutamine
• Phenol Red• Sodium Pyruvate
• HEPES
DMEM is unique from other media as it contains 4 times the concentration of amino acids and vitamins than the original Eagle's Minimal Essential Medium. DMEM was originally formulated with low glucose (1 g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate.
DMEM contains no proteins, lipids, or growth factors. Therefore, DMEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM uses a sodium bicarbonate buffer system (3.7 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Gibco® 'BASIC' products offer value-minded scientists liquid cell culture media and buffer solutions at a significant price savings. As part of Life Technologies’ commitment to quality, this select range of Research Use Only products is continually tested to ensure performance and customer satisfaction.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Unit Size500 mL
Contents & Storage
Storage conditions: 2-8° C. Protect from lightShipping conditions: AmbientShelf life: 12 months from date of manufacture