Glycerol, 99+%, Certified AR for Analysis, Fisher Chemical™
Glycerol, 99+%, Certified AR for Analysis, Fisher Chemical™
Glycerol, 99+%, Certified AR for Analysis, Fisher Chemical™
Fisher Chemical™

Glycerol, 99+%, Certified AR for Analysis, Fisher Chemical™

C3H8O3, CAS Number-56-81-5, propanetriol, trihydroxypropane, glycyl alcohol, glycerol, glyceritol, glycerin, glycerine, osmoglyn, 1,2,3-propanetriol, 1,2,3-trihydroxypropane, 2.5L, 290 deg.C, CHEBI:17754, Undesignated, 92.09g/mol, PEDCQBHIVMGVHV-UHFFFAOYSA-N, propane-1,2,3-triol, 18 deg.C, 4722 | CAS: 56-81-5 | C3H8O3 | 92.09 g/mol
Quantity:
500 mL
1 L
2.5 L
Packaging:
HDPE plastic bottle
Catalog number G-0650-15
also known as G/0650/15
Price (JPY)
-
Quantity:
1 L
Packaging:
HDPE plastic bottle
Request bulk or custom format
Specifications
CAS56-81-5
ChEBICHEBI:17754
Formula Weight92.09g/mol
IUPAC Namepropane-1,2,3-triol
InChI KeyPEDCQBHIVMGVHV-UHFFFAOYSA-N
View more
C3H8O3, CAS Number-56-81-5, glycerol, propanetriol, glycyl alcohol, glycerin, 1,2,3-trihydroxypropane, glycerine, 1,2,3-propanetriol, trihydroxypropane, osmoglyn, glyceritol, 1L, 290 deg.C, CHEBI:17754, Undesignated, 92.09g/mol, PEDCQBHIVMGVHV-UHFFFAOYSA-N, propane-1,2,3-triol, 18 deg.C, 4722