(Pentafluoroethyl)trimethylsilane, 97%
(Pentafluoroethyl)trimethylsilane, 97%
(Pentafluoroethyl)trimethylsilane, 97%
Thermo Scientific Chemicals

(Pentafluoroethyl)trimethylsilane, 97%

CAS: 124898-13-1 | C5H9F5Si | 192.204 g/mol
Quantity:
1 g
5 g
Catalog number H25775.06
also known as H25775-06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS124898-13-1
IUPAC Nametrimethyl(1,1,2,2,2-pentafluoroethyl)silane
Molecular FormulaC5H9F5Si
InChI KeyMTPVUVINMAGMJL-UHFFFAOYSA-N
SMILESC[Si](C)(C)C(F)(F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.3230-1.3280 @ 20?C
(Pentafluoroethyl)trimethylsilane acts as a silicon-based fluorinating reagent for trifluoromethylation of carbonyls and ethylene diamines. It is also employed as a precursor for metal-mediated trifluoromethylation of aromatic, heteroaromatic and vinyl substrates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Pentafluoroethyl)trimethylsilane acts as a silicon-based fluorinating reagent for trifluoromethylation of carbonyls and ethylene diamines. It is also employed as a precursor for metal-mediated trifluoromethylation of aromatic, heteroaromatic and vinyl substrates.

Solubility
Miscible with tetrahydrofuran.

Notes
Moisture sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Jin, G.; Zhang, X.; Fu, D.; Dai, W.; Cao, S. Mild and metal-free trifluoromethylation and pentafluoroethylation of gem-difluoroalkenes with TMSCF3 and TMSCF2CF3. Tetrahedron 2015, 71 (41), 7892-7899.
  2. O’Duill, M.; Dubost, E.; Pfeifer, L.; Gouverneur, V. Cross-Coupling of [2-Aryl-1,1,2,2-tetrafluoroethyl](trimethyl)silanes with Aryl Halides. Org. Lett. 2015, 17 (14), 3466-3469.