4-Amino-3-fluorobenzoic acid, 98%, Thermo Scientific Chemicals
4-Amino-3-fluorobenzoic acid, 98%, Thermo Scientific Chemicals
4-Amino-3-fluorobenzoic acid, 98%, Thermo Scientific Chemicals
4-Amino-3-fluorobenzoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Amino-3-fluorobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog NumberQuantity
H26092.03
also known as H26092-03
1 g
Catalog number H26092.03
also known as H26092-03
Price (JPY)
-
Request A Quote
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS455-87-8
IUPAC Name4-amino-3-fluorobenzoic acid
Molecular FormulaC7H6FNO2
InChI KeyJSKXHTHMCCDEGD-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1F)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (Silylated GC)≥97.5%
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Melting Point (clear melt)213-217?C (≤2?C range)
Appearance (Color)White to cream or pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only