3-Fluoro-4-hydroxybenzonitrile, 98%, Thermo Scientific Chemicals
3-Fluoro-4-hydroxybenzonitrile, 98%, Thermo Scientific Chemicals
3-Fluoro-4-hydroxybenzonitrile, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Fluoro-4-hydroxybenzonitrile, 98%, Thermo Scientific Chemicals

Catalog NumberQuantity
H26107.03
also known as H26107-03
1 g
Catalog number H26107.03
also known as H26107-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS405-04-9
IUPAC Name3-fluoro-4-hydroxybenzonitrile
Molecular FormulaC7H4FNO
InChI KeyDPSSSDFTLVUJDH-UHFFFAOYSA-N
SMILESOC1=C(F)C=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)131.0-137.0?C
Appearance (Color)White to pale cream
Assay (GC)≥97.5%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only