4-(Trifluoromethoxy)phenylhydrazine hydrochloride, 98%
4-(Trifluoromethoxy)phenylhydrazine hydrochloride, 98%
4-(Trifluoromethoxy)phenylhydrazine hydrochloride, 98%
Thermo Scientific Chemicals

4-(Trifluoromethoxy)phenylhydrazine hydrochloride, 98%

CAS: 133115-72-7 | C7H8ClF3N2O | 228.60 g/mol
Quantity:
1 g
5 g
25 g
Catalog number H26180.14
also known as H26180-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS133115-72-7
IUPAC Name[4-(trifluoromethoxy)phenyl]hydrazine hydrochloride
Molecular FormulaC7H8ClF3N2O
InChI KeyKQXZVSQCMVKMBK-UHFFFAOYSA-N
SMILESCl.NNC1=CC=C(OC(F)(F)F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream to pale brown
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
Assay (HPLC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only