Dichloro[1,1'-bis(diphenylphosphino)ferrocene]nickel(II), 99%, Thermo Scientific Chemicals
Dichloro[1,1'-bis(diphenylphosphino)ferrocene]nickel(II), 99%, Thermo Scientific Chemicals
Dichloro[1,1'-bis(diphenylphosphino)ferrocene]nickel(II), 99%, Thermo Scientific Chemicals
Dichloro[1,1'-bis(diphenylphosphino)ferrocene]nickel(II), 99%, Thermo Scientific Chemicals
Dichloro[1,1'-bis(diphenylphosphino)ferrocene]nickel(II), 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dichloro[1,1'-bis(diphenylphosphino)ferrocene]nickel(II), 99%, Thermo Scientific Chemicals

Quantity:
1 g
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number H26895.14
also known as H26895-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS67292-34-6
IUPAC NameDichloro[1,1'-bis(diphenylphosphino)ferrocene]nickel(II)
Molecular FormulaC34H28Cl2FeNiP2
SMILES[Fe].Cl[Ni]Cl.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol)683.98
View more
SpecificationsSpecification SheetSpecification Sheet
Elemental AnalysisCarbon C: 58.81 - 60.60% (Theory 59.70%)
FormCrystalline powder
Assay (unspecified)≥98.5% based on Carbon EA
Elemental AnalysisHydrogen H: 4.03 - 4.21% (Theory 4.13%)
Appearance (Color)Dark green
diphosphine 1,1'-bis(diphenylphosphino)ferrocene (dppf) are used in transition metal-catalyzed reactions to synthesize pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
diphosphine 1,1′-bis(diphenylphosphino)ferrocene (dppf) are used in transition metal-catalyzed reactions to synthesize pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents
RUO – Research Use Only

General References:

  1. Snegur,; L.V.; Simenel,; A. A.; Rodionov,; A. N.; Boev, V. I. Ferrocene modification of organic compounds for medicinal applications. Russ. Chem. Bull. 2014 , 63 26-36.
  2. authors. title . Journal name. year of publication , volume issue , pg no.