Tris(cyclopentadienyl)terbium(III), 99% (99.9%-Tb) (REO)
Tris(cyclopentadienyl)terbium(III), 99% (99.9%-Tb) (REO)
Tris(cyclopentadienyl)terbium(III), 99% (99.9%-Tb) (REO)
Thermo Scientific Chemicals

Tris(cyclopentadienyl)terbium(III), 99% (99.9%-Tb) (REO)

CAS: 1272-25-9 | C15H15Tb | 354.21 g/mol
Quantity:
1 g
5 g
Catalog number H27238.03
also known as H27238-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS1272-25-9
IUPAC Nametris(cyclopenta-2,4-dien-1-yl)terbium
Molecular FormulaC15H15Tb
InChI KeyFMNCAPFXHVPBJH-UHFFFAOYSA-N
SMILESC1=CC(C=C1)[Tb](C1C=CC=C1)C1C=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only