1-Iodo-4-n-pentylbenzene, 98%
1-Iodo-4-n-pentylbenzene, 98%
1-Iodo-4-n-pentylbenzene, 98%
1-Iodo-4-n-pentylbenzene, 98%
Thermo Scientific Chemicals

1-Iodo-4-n-pentylbenzene, 98%

CAS: 85017-60-3 | C11H15I | 274.145 g/mol
Quantity:
1 g
10 g
Catalog number H27356.09
also known as H27356-09
Price (JPY)
-
Request A Quote
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS85017-60-3
IUPAC Name1-iodo-4-pentylbenzene
Molecular FormulaC11H15I
InChI KeyMTRMOYSZRPLAOL-UHFFFAOYSA-N
SMILESCCCCCC1=CC=C(I)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
Refractive Index1.5570-1.5610 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only