4-(2-Thienyl)aniline, 97%
4-(2-Thienyl)aniline, 97%
4-(2-Thienyl)aniline, 97%
4-(2-Thienyl)aniline, 97%
Thermo Scientific Chemicals

4-(2-Thienyl)aniline, 97%

CAS: 70010-48-9 | C10H9NS | 175.249 g/mol
Catalog NumberQuantity
H27753.03
also known as H27753-03
1 g
Catalog number H27753.03
also known as H27753-03
Price (JPY)
-
Request A Quote
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS70010-48-9
IUPAC Name4-(thiophen-2-yl)aniline
Molecular FormulaC10H9NS
InChI KeyWPNWHSQJRALLBJ-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1)C1=CC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)Brown
Assay (GC)≥96.0%
Melting Point (clear melt)71.0-80.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only