Ethyl 2-chlorooxazole-4-carboxylate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 2-chlorooxazole-4-carboxylate, 95%, Thermo Scientific Chemicals

Catalog NumberQuantity
H31521.03
also known as H31521-03
1 g
Catalog number H31521.03
also known as H31521-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS460081-18-9
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (GC)≥94.0%
Appearance (Color)White to cream to orange or pink
Melting Point (clear melt)44.5-53.5°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only