3-Amino-2,6-dichloropyridine, 97%
3-Amino-2,6-dichloropyridine, 97%
3-Amino-2,6-dichloropyridine, 97%
3-Amino-2,6-dichloropyridine, 97%
Thermo Scientific Chemicals

3-Amino-2,6-dichloropyridine, 97%

CAS: 62476-56-6 | C5H4Cl2N2 | 163.00 g/mol
Quantity:
5 g
25 g
Catalog number H31598.06
also known as H31598-06
Price (JPY)
-
Request A Quote
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS62476-56-6
IUPAC Name2,6-dichloropyridin-3-amine
Molecular FormulaC5H4Cl2N2
InChI KeyMJVZSRZTBDMYLX-UHFFFAOYSA-N
SMILESNC1=CC=C(Cl)N=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)117.0-126.0?C
Appearance (Color)White to yellow to brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only