2-Bromo-4-(trifluoromethyl)benzenesulfonamide, 97%, Thermo Scientific Chemicals
2-Bromo-4-(trifluoromethyl)benzenesulfonamide, 97%, Thermo Scientific Chemicals
2-Bromo-4-(trifluoromethyl)benzenesulfonamide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromo-4-(trifluoromethyl)benzenesulfonamide, 97%, Thermo Scientific Chemicals

Quantity:
1 g
5 g
Catalog number H31649.03
also known as H31649-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS351003-63-9
IUPAC Name2-bromo-4-(trifluoromethyl)benzene-1-sulfonamide
Molecular FormulaC7H5BrF3NO2S
InChI KeyIJUIBMWZUXWVLI-UHFFFAOYSA-N
SMILESNS(=O)(=O)C1=C(Br)C=C(C=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (GC)≥96.0%
Appearance (Color)Pale yellow
Melting Point (clear melt)157.0-166.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only