4-Bromo-2-(trifluoromethyl)benzoic acid, 98%
4-Bromo-2-(trifluoromethyl)benzoic acid, 98%
4-Bromo-2-(trifluoromethyl)benzoic acid, 98%
Thermo Scientific Chemicals

4-Bromo-2-(trifluoromethyl)benzoic acid, 98%

CAS: 320-31-0 | C8H4BrF3O2 | 269.02 g/mol
Quantity:
1 g
5 g
Catalog number H31656.03
also known as H31656-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS320-31-0
IUPAC Name4-bromo-2-(trifluoromethyl)benzoic acid
Molecular FormulaC8H4BrF3O2
InChI KeyJXHWAPDBDXPBEQ-UHFFFAOYSA-N
SMILESOC(=O)C1=C(C=C(Br)C=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Melting Point (clear melt)119.5-125.5?C
FormCrystals or powder or crystalline powder
Appearance (Color)White to pale cream
Assay (Silylated GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only