Methyl 2-hydroxy-5,6,7,8-tetrahydronaphthalene-1-carboxylate, 97%, Thermo Scientific Chemicals
Methyl 2-hydroxy-5,6,7,8-tetrahydronaphthalene-1-carboxylate, 97%, Thermo Scientific Chemicals
Methyl 2-hydroxy-5,6,7,8-tetrahydronaphthalene-1-carboxylate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 2-hydroxy-5,6,7,8-tetrahydronaphthalene-1-carboxylate, 97%, Thermo Scientific Chemicals

Catalog NumberQuantity
H31668.03
also known as H31668-03
1 g
Catalog number H31668.03
also known as H31668-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS59604-96-5
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)33.0-43.0?C
Appearance (Color)Pale cream
Assay (GC)≥96.0%
FormPowder or fused solid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only