5-Methyl-3-phenylisoxazole-4-carboxaldehyde, 97%
5-Methyl-3-phenylisoxazole-4-carboxaldehyde, 97%
5-Methyl-3-phenylisoxazole-4-carboxaldehyde, 97%
Thermo Scientific Chemicals

5-Methyl-3-phenylisoxazole-4-carboxaldehyde, 97%

CAS: 87967-95-1 | C11H9NO2 | 187.20 g/mol
Catalog NumberQuantity
H32129.03
also known as H32129-03
1 g
Catalog number H32129.03
also known as H32129-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS87967-95-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (GC)≥96.0%
Melting Point (clear melt)51.0-57.0?C
FormCrystals or powder or crystalline powder
Free acid (titration)≤2.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only