2-Methylhexanoyl chloride, 97%
2-Methylhexanoyl chloride, 97%
2-Methylhexanoyl chloride, 97%
2-Methylhexanoyl chloride, 97%
Thermo Scientific Chemicals

2-Methylhexanoyl chloride, 97%

CAS: 41693-47-4 | C7H13ClO | 148.63 g/mol
Quantity:
1 g
10 g
Catalog number H32829.03
also known as H32829-03
Price (JPY)
-
Request A Quote
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS41693-47-4
IUPAC Name2-methylhexanoyl chloride
Molecular FormulaC7H13ClO
InChI KeyWKEBLSOTVYFYKY-UHFFFAOYNA-N
SMILESCCCCC(C)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)Clear colorless
Refractive Index1.4270-1.4320 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only