(Morpholinium-4-ylmethyl)trifluoroborate internal salt, 95%
(Morpholinium-4-ylmethyl)trifluoroborate internal salt, 95%
(Morpholinium-4-ylmethyl)trifluoroborate internal salt, 95%
(Morpholinium-4-ylmethyl)trifluoroborate internal salt, 95%
Thermo Scientific Chemicals

(Morpholinium-4-ylmethyl)trifluoroborate internal salt, 95%

CAS: 1268340-94-8 | C5H11BF3NO | 168.954 g/mol
Catalog NumberQuantity
H33149.06
also known as H33149-06
5 g
Catalog number H33149.06
also known as H33149-06
Price (JPY)
-
Request A Quote
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS1268340-94-8
IUPAC Nametrifluoro[(morpholin-4-ium-4-yl)methyl]boranuide
Molecular FormulaC5H11BF3NO
InChI KeyFQKLIGKQQGNVHW-UHFFFAOYSA-O
SMILESF[B-](F)(F)C[NH+]1CCOCC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Proton NMRConforms to structure
Carbon-13 NMRConforms to structure
Appearance (Color)White to cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only