3-(4-Nitrophenyl)propionic acid, 96%
3-(4-Nitrophenyl)propionic acid, 96%
3-(4-Nitrophenyl)propionic acid, 96%
Thermo Scientific Chemicals

3-(4-Nitrophenyl)propionic acid, 96%

CAS: 16642-79-8 | C9H9NO4 | 195.17 g/mol
Quantity:
10 g
50 g
Catalog number H33663.18
also known as H33663-18
Price (JPY)
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS16642-79-8
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)161.0-170.0?C
Appearance (Color)White to yellow
FormCrystals or powder or crystalline powder
Assay (HPLC)≥97.5%
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed containers. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Meihua Liu; Guangyu Shen; Weijian Xu; and Guoli Shen. Synthesis of a novel aromatic-aliphatic hyperbranched polyamide and its application in piezoelectric immunosensors. Polymer International. 200756 (11), 1432-1439.
  2. Javier Magano; Douglas Farrand; Jeffrey P. Haase; Michael Lovdahl; Mark T. Maloney; Mark J. Pozzo; John J. Teixeira; David C. Whritenour; John Rizzod, David Tumelty; Abhijit Bhat; Curt Bradshaw.Synthesis of a novel analytical reagent for the determination of active sites for conjugation on a catalytic aldolase monoclonal antibody. Tetrahedron Letters. 201253 (11), 1385-1389.