12-Bromododecylphosphonic acid, 95%
12-Bromododecylphosphonic acid, 95%
12-Bromododecylphosphonic acid, 95%
Thermo Scientific Chemicals

12-Bromododecylphosphonic acid, 95%

CAS: 202920-07-8 | C12H26BrO3P | 329.22 g/mol
Quantity:
250 mg
1 g
Catalog number H33714.03
also known as H33714-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS202920-07-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥94.0 to ≤106.0%
Melting Point (clear melt)87.5-96.5?C
FormPowder
Employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as a intermediate for pharmaceutical.

Notes
Store in cool, dry place in tightly closed containers. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Tom Hauffman; Orlin Blajiev; Johan Snauwaert; Chris van Haesendonck; Annick Hubin; and Herman Terryn. Study of the Self-Assembling of n-Octylphosphonic Acid Layers on Aluminum Oxide. Langmuir. 200824 (23), 13450-13456.
  2. Charlene Presti; Johan G. Alauzun; Danielle Laurencin; and P. Hubert Mutin. Surface Functionalization of Detonation Nanodiamonds by Phosphonic Dichloride Derivatives. Langmuir. 201430 (30), 9239-9245.