2,2'-bipyridine-5,5'-dicarboxylic acid, 97%, Thermo Scientific Chemicals
2,2'-bipyridine-5,5'-dicarboxylic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2'-bipyridine-5,5'-dicarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog NumberQuantity
H36088.03
also known as H36088-03
1 g
Catalog number H36088.03
also known as H36088-03
Price (JPY)
-
Request A Quote
Quantity:
1 g
Request bulk or custom format
Specifications
Melting Point365°C
Percent Purity97%
Physical FormSolid
Quantity1 g
Unit SizeEach
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.