4-Chloro-8-methoxy-2-methylquinoline, 95%
4-Chloro-8-methoxy-2-methylquinoline, 95%
4-Chloro-8-methoxy-2-methylquinoline, 95%
4-Chloro-8-methoxy-2-methylquinoline, 95%
Thermo Scientific Chemicals

4-Chloro-8-methoxy-2-methylquinoline, 95%

CAS: 64951-58-2 | C11H10ClNO | 207.66 g/mol
Quantity:
1 g
5 g
Catalog number H50559.06
also known as H50559-06
Price (JPY)
-
Request A Quote
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS64951-58-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow or pale purple to brown
FormCrystals or powder or crystalline powder
Assay (HPLC)≥94.0%
Proton NMRConforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only