1-Chloro-5-triethylsilyl-4-pentyne, 97%
1-Chloro-5-triethylsilyl-4-pentyne, 97%
1-Chloro-5-triethylsilyl-4-pentyne, 97%
Thermo Scientific Chemicals

1-Chloro-5-triethylsilyl-4-pentyne, 97%

CAS: 174125-30-5 | C11H21ClSi | 216.82 g/mol
Quantity:
5 g
25 g
Catalog number H53515.14
also known as H53515-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Specifications
CAS174125-30-5
Chemical Name or Material1-Chloro-5-triethylsilyl-4-pentyne
Formula Weight216.83
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only